10-Undecynoic acid ≥98.0%

Supplier: TCI America

U0054-5G U0054-1G
TCU0054-005GEA 151.02 CAD
TCU0054-005G TCU0054-001G
10-Undecynoic acid ≥98.0%
10-Undecynoic acid

CAS Number: 2777-65-3
MDL Number: MFCD00014389
Molecular Formula: C11H18O2
Molecular Weight: 182.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 180
Melting point (°C): 43
Flash Point (°C): 110

Formula: C₁₁H₁₈O₂
Boiling Pt: 180 °C
Melting Pt: 43 °C
Flash Pt: 110 °C
MDL Number: MFCD00014389
CAS Number: 2777-65-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c421f51673464ea7800a9b7b7dd9742c52b3594&vid=3c421f51673464ea7800a9b7b7dd9742c52b3594&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR