Undecanophenone ≥98.0%
Supplier: TCI America
U0053-500G
U0053-25G
TCU0053-500GEA
1485.99
CAD
TCU0053-500G
TCU0053-025G
Undecanophenone ≥98.0%
Undecanophenone
CAS Number: 4433-30-1
MDL Number: MFCD00051548
Molecular Formula: C17H26O
Molecular Weight: 246.39
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Freezing point (°C): 28
Other Product Names / Synonyms: Decyl Phenyl Ketone
Formula:
C₁₇H₂₆O |
MDL Number:
MFCD00051548 CAS Number: 4433-30-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...