Luteolin ≥98.0% (by HPLC)
Supplier: TCI America
T2682-1G
T2682-5G
TCT2682-1GEA
173.52
CAD
TCT2682-1G
TCT2682-5G
Luteolin ≥98.0% (by HPLC)
Luteolin
CAS Number: 491-70-3
MDL Number: MFCD00017309
Molecular Formula: C15H10O6
Molecular Weight: 286.24
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 330
Storage Temperature: 0-10°C
Product Categories: Bioscience, Antioxidants, Aromatic Polyketides, Polyphenols (Phytochemicals)
Other Product Names / Synonyms: Luteolin
Formula:
C₁₅H₁₀O₆ Melting Pt: 330 °C |
MDL Number:
MFCD00017309 CAS Number: 491-70-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...