2,4,6-Trimethoxybenzaldehyde ≥98.0%
Supplier: TCI America
T2651-25G
T2651-5G
TCT2651-25GEA
225.94
CAD
TCT2651-25G
TCT2651-5G
2,4,6-Trimethoxybenzaldehyde ≥98.0%
2,4,6-Trimethoxybenzaldehyde
CAS Number: 830-79-5
MDL Number: MFCD00003313
Molecular Formula: C10H12O4
Molecular Weight: 196.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 120
Formula:
C₁₀H₁₂O₄ MW: 196.2 g/mol Melting Pt: 120 °C |
MDL Number:
MFCD00003313 CAS Number: 830-79-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...