3,4,5-Trihydroxybenzaldehyde ≥98.0% (by GC, titration analysis)

Supplier: TCI America

T2650-5G T2650-1G
TCT2650-5GEA 925.56 CAD
TCT2650-5G TCT2650-1G
3,4,5-Trihydroxybenzaldehyde ≥98.0% (by GC, titration analysis)
3,4,5-Trihydroxybenzaldehyde

Form: Crystal
Color: White
Melting point (°C): 212


Product Categories: Bioscience, Antioxidants
Other Product Names / Synonyms: Pyrogallol-5-carboxaldehyde

Formula: C₇H₆O₄
Melting Pt: 212 °C
MDL Number: MFCD00003371
CAS Number: 13677-79-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca14924719201197601ddcfb5924d6ed12c1c05e&vid=ca14924719201197601ddcfb5924d6ed12c1c05e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR