2,4,5-Trimethoxybenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
T2627-25G
T2627-5G
TCT2627-25GEA
85.32
CAD
TCT2627-25G
TCT2627-5G
2,4,5-Trimethoxybenzoic acid ≥98.0% (by GC, titration analysis)
2,4,5-Trimethoxybenzoic acid
CAS Number: 490-64-2
MDL Number: MFCD00002435
Molecular Formula: C10H12O5
Molecular Weight: 212.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 300
Melting point (°C): 144
Lambda max.: 226 nm (EtOH)
Formula:
C₁₀H₁₂O₅ MW: 212.2 g/mol Boiling Pt: 300 °C Melting Pt: 144 °C |
MDL Number:
MFCD00002435 CAS Number: 490-64-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...