1-(2-Tetrahydrofuroyl)piperazine ≥97.0%

Supplier: TCI America

Synonyms: (Piperazin-1-yl)(tetrahydrofuran-2-yl)methanone

T2617-25G T2617-5G
TCT2617-25GEA 159.58 CAD
TCT2617-25G TCT2617-5G
1-(2-Tetrahydrofuroyl)piperazine ≥97.0%
1-(2-Tetrahydrofuroyl)piperazine

CAS Number: 63074-07-7
MDL Number: MFCD00190247
Molecular Formula: C9H16N2O2
Molecular Weight: 184.24
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Pale Yellow
Boiling point (°C): 125
Specific Gravity (20/20): 1.17


Other Product Names / Synonyms: 1-[(Tetrahydrofuran-2-yl)carbonyl]piperazine

Formula: C₉H₁₆N₂O₂
MW: 184.24 g/mol
Boiling Pt: 125 °C
Density: 1.17 (20 °C)
MDL Number: MFCD00190247
CAS Number: 63074-07-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91c01e125f0661116ee7dfe223f819e1456990b0&vid=91c01e125f0661116ee7dfe223f819e1456990b0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR