2-(Trifluoromethyl)cinnamic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
T2608-25G
T2608-5G
TCT2608-25GEA
301.78
CAD
TCT2608-25G
TCT2608-5G
2-(Trifluoromethyl)cinnamic acid ≥98.0% (by GC, titration analysis)
2-(Trifluoromethyl)cinnamic acid
CAS Number: 2062-25-1
MDL Number: MFCD00004381
Molecular Formula: C10H7F3O2
Molecular Weight: 216.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 204
Formula:
C₁₀H₇F₃O₂ Melting Pt: 204 °C |
MDL Number:
MFCD00004381 CAS Number: 2062-25-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...