2-(Trifluoromethoxy)benzenesulfonamide ≥98.0% (by GC, titration analysis)
Supplier: TCI America
T2604-5G
T2604-25G
TCT2604-5GEA
37.92
CAD
TCT2604-5G
TCT2604-25G
2-(Trifluoromethoxy)benzenesulfonamide ≥98.0% (by GC, titration analysis)
2-(Trifluoromethoxy)benzenesulphonamide
CAS Number: 37526-59-3
MDL Number: MFCD01320751
Molecular Formula: C7H6F3NO3S
Molecular Weight: 241.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 188
Formula:
C₇H₆F₃NO₃S Melting Pt: 188 °C |
MDL Number:
MFCD01320751 CAS Number: 37526-59-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...