1-Iodo-3,3,3-trifluoropropane ≥98.0%
Supplier: TCI America
Synonyms:
1,1,1-Trifluoro-3-iodopropane, 3-Iodo-1,1,1-trifluoropropane
T2588-25G
T2588-5G
TCT2588-25GEA
192.76
CAD
TCT2588-25G
TCT2588-5G
1-Iodo-3,3,3-trifluoropropane ≥98.0%
1-Iodo-3,3,3-trifluoropropane
CAS Number: 460-37-7
MDL Number: MFCD00038531
Molecular Formula: C3H4F3I
Molecular Weight: 223.96
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 90
Specific Gravity (20/20): 1.97
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3,3,3-Trifluoropropyl Iodide
Formula:
C₃H₄F₃I MW: 223.96 g/mol Boiling Pt: 90 °C Density: 1.97 (20 °C) |
MDL Number:
MFCD00038531 CAS Number: 460-37-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...