3,3',5,5'-Tetramethylbenzidine (TMB) ≥98.0% (by HPLC, titration analysis) for biochemical research

Supplier: TCI America

Synonyms: TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine

T2573-5G T2573-1G
TCT2573-5GEA 265.28 CAD
TCT2573-5G TCT2573-1G
3,3',5,5'-Tetramethylbenzidine (TMB) ≥98.0% (by HPLC, titration analysis) for biochemical research
3,3',5,5'-Tetramethylbenzidine (TMB)

[for Biochemical Research] CAS Number: 54827-17-7 MDL Number: MFCD00007748 Molecular Formula: C16H20N2 Molecular Weight: 240.35 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal Color: White Melting point (°C): 170


Product Categories: Bioscience, Peroxidase Substrates

Formula: C₁₆H₂₀N₂
MW: 240.35 g/mol
Boiling Pt: 402
Melting Pt: 170 °C
MDL Number: MFCD00007748
CAS Number: 54827-17-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b7cf5c35f6a6b91e176f58848dee9c415f33f98&vid=8d340dc7f0e14e1c07c30435e7751fbb7ae3c0a3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR