Triisopropoxyvanadium(V) oxide ≥97.0%

Supplier: TCI America

Synonyms: Vanadium(V) Triisopropoxide Oxide, Vanadium (V) tri-i-propoxy oxide, Vanadium(V) Oxytriisopropoxide, VTIP

T2548-25G T2548-5G
TCT2548-25GEA 243.32 CAD
TCT2548-25G TCT2548-5G
Triisopropoxyvanadium(V) oxide ≥97.0%
Triisopropoxyvanadium(V) oxide

CAS Number: 5588-84-1
MDL Number: MFCD00015017
Molecular Formula: C9H21O4V
Molecular Weight: 244.20
Purity/Analysis Method: >97.0% (T)
Form: Clear Liquid
Color: Pale Yellow
Boiling point (°C): 102
Melting point (°C): -2
Flash Point (°C): 70
Specific Gravity (20/20): 1.04


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts
Other Product Names / Synonyms: Vanadium(V) Oxytriisopropoxide, Vanadium(V) Triisopropoxide Oxide

Formula: C₉H₂₁O₄V
MW: 244.20 g/mol
Boiling Pt: 102 °C
Melting Pt: –2 °C
Density: 1.04 (20 °C)
Flash Pt: 70 °C
MDL Number: MFCD00015017
CAS Number: 5588-84-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=063e9a889304572b82944070af1fec61d30388e0&vid=063e9a889304572b82944070af1fec61d30388e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR