1,1,3,3-Tetrabutylurea ≥98.0%

Supplier: TCI America

T2537-5G T2537-25G
TCT2537-5GEA 150.42 CAD
TCT2537-5G TCT2537-25G
1,1,3,3-Tetrabutylurea ≥98.0%
1,1,3,3-Tetrabutylurea

CAS Number: 4559-86-8
MDL Number: MFCD00042901
Molecular Formula: C17H36N2O
Molecular Weight: 284.49
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 163
Melting point (°C): -60
Flash Point (°C): 93
Specific Gravity (20/20): 0.88

Formula: C₁₇H₃₆N₂O
MW: 284.49 g/mol
Boiling Pt: 163 °C
Melting Pt: –60 °C
Density: 0.88 (20 °C)
Flash Pt: 93 °C
MDL Number: MFCD00042901
CAS Number: 4559-86-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2790647d299729a456b686655909fafa53774cc2&vid=2790647d299729a456b686655909fafa53774cc2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR