Tween® 80 (Polysorbate) for biochemical research
Supplier: TCI America
T2533-25G
T2533-5G
TCT2533-25GEA
91.32
CAD
TCT2533-25G
TCT2533-5G
Tween® 80 (Polysorbate) for biochemical research
Tween® 80 (Polysorbate)
[for Biochemical Research]
CAS Number: 9005-65-6
MDL Number: MFCD00082107
Form: Clear Liquid
Color: Slightly Pale Yellow
Melting point (°C): -25
Flash Point (°C): 295
Specific Gravity (20/20): 1.08
Other Product Names / Synonyms: Polyoxyethylene Sorbitan Monooleate
Melting Pt:
–25 °C Density: 1.08 (20 °C) Flash Pt: 295 °C |
MDL Number:
MFCD00082107 CAS Number: 9005-65-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...