Tween® 60 (Polysorbate) for biochemical research
Supplier: TCI America
T2532-25G
T2532-5G
TCT2532-25GEA
55.3
CAD
TCT2532-25G
TCT2532-5G
Tween® 60 (Polysorbate) for biochemical research
Tween® 60 (Polysorbate)
[for Biochemical Research]
CAS Number: 9005-67-8
MDL Number: MFCD01780110
Form: Crystal
Color: Pale Yellow
Flash Point (°C): 277
Freezing point (°C): 23
Other Product Names / Synonyms: Polyoxyethylene Sorbitan Monostearate
Formula:
C₆₄H₁₂₆O₂₆ Flash Pt: 277 °C |
MDL Number:
MFCD01780110 CAS Number: 9005-67-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...