Tween® 40 (Polysorbate) for biochemical research

Supplier: TCI America

T2531-25G T2531-5G
TCT2531-25GEA 61.62 CAD
TCT2531-25G TCT2531-5G
Tween® 40 (Polysorbate) for biochemical research
Tween® 40 (Polysorbate)

[for Biochemical Research]
CAS Number: 9005-66-7
MDL Number: MFCD00165345
Form: Clear Liquid
Color: Pale Yellow
Melting point (°C): 20
Flash Point (°C): 268
Specific Gravity (20/20): 1.10


Other Product Names / Synonyms: Polyoxyethylene Sorbitan Monopalmitate

Formula: C₆₂H₁₂₂O₂₆
Melting Pt: 20 °C
Density: 1.10 (20 °C)
Flash Pt: 268 °C
MDL Number: MFCD00165345
CAS Number: 9005-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR