Tween® 20 (Polysorbate) for biochemical research

Supplier: TCI America

Synonyms: Polyoxyethylene sorbitan monolaurate

T2530-25G T2530-5G
TCT2530-25GEA 70.97 CAD
TCT2530-25G TCT2530-5G
Tween® 20 (Polysorbate) for biochemical research
Tween® 20 (Polysorbate)

[for Biochemical Research]
CAS Number: 9005-64-5
MDL Number: MFCD00165986
Form: Clear Liquid
Color: Yellow
Melting point (°C): -14
Flash Point (°C): 297
Specific Gravity (20/20): 1.11


Other Product Names / Synonyms: Polyoxyethylene Sorbitan Monolaurate

Formula: C₅₈H₁₁₄O₂₆
MW: 1227.53 g/mol
Melting Pt: –14 °C
Density: 1.11 (20 °C)
Flash Pt: 297 °C
MDL Number: MFCD00165986
CAS Number: 9005-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR