1H,1H,2H,2H-Tridecafluoro-1-octanol ≥98.0%

Supplier: TCI America

Synonyms: 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctan-1-ol, 1H,1H,2H,2H-Perfluoro-1-octanol, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-1-octanol, 1H,1H,2H,2H-Perfluorooctan-1-ol, 1H,1H,2H,2H-Perfluorooctanol

T2528-25G T2528-5G
TCT2528-25GEA 165.9 CAD
TCT2528-25G TCT2528-5G
1H,1H,2H,2H-Tridecafluoro-1-octanol ≥98.0%
1H,1H,2H,2H-Tridecafluoro-1-octanol

CAS Number: 647-42-7
MDL Number: MFCD00042143
Molecular Formula: C8H5F13O
Molecular Weight: 364.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 87
Flash Point (°C): 91
Specific Gravity (20/20): 1.69


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks, Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: 2-(Perfluorohexyl)ethanol

Formula: C₈H₅F₁₃O
MW: 364.11 g/mol
Boiling Pt: 87 °C
Density: 1.69 (20 °C)
Flash Pt: 91 °C
MDL Number: MFCD00042143
CAS Number: 647-42-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dbc7d11d291175188f71c73dbea523d3153dffa2&vid=dbc7d11d291175188f71c73dbea523d3153dffa2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR