2,4,5-Trichloropyrimidine ≥98.0%
Supplier: TCI America
T2512-5G
T2512-25G
TCT2512-5GEA
128.61
CAD
TCT2512-5G
TCT2512-25G
2,4,5-Trichloropyrimidine ≥98.0%
2,4,5-Trichloropyrimidine
CAS Number: 5750-76-5
MDL Number: MFCD03788200
Molecular Formula: C4HCl3N2
Molecular Weight: 183.42
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 96
Flash Point (°C): 110
Specific Gravity (20/20): 1.61
Formula:
C₄HCl₃N₂ MW: 183.42 g/mol Boiling Pt: 96 °C Density: 1.61 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD03788200 CAS Number: 5750-76-5 UN: 3334 ADR: 9, |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...