4-(Trifluoromethyl)cinnamic acid ≥98.0%

Supplier: TCI America

Synonyms: trans-4-(Trifluoromethyl)cinnamic acid

T2499-5G T2499-1G T2499-25G
TCT2499-5GEA 110.6 CAD
TCT2499-5G TCT2499-1G TCT2499-25G
4-(Trifluoromethyl)cinnamic acid ≥98.0%
4-(Trifluoromethyl)cinnamic acid

CAS Number: 16642-92-5
MDL Number: MFCD00002696
Molecular Formula: C10H7F3O2
Molecular Weight: 216.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 231

Formula: C₁₀H₇F₃O₂
MW: 216.16 g/mol
Melting Pt: 231 °C
MDL Number: MFCD00002696
CAS Number: 2062-26-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1a83b9fb1777b29cb28ce9137b5dcc4e1c8e49a&vid=c1a83b9fb1777b29cb28ce9137b5dcc4e1c8e49a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR