2,4,6-Trivinylboroxin pyridine complex ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Trivinylcyclotriboroxan - Pyridine Complex
T2498-1G
T2498-5G
TCT2498-1GEA
148.52
CAD
TCT2498-1G
TCT2498-5G
2,4,6-Trivinylboroxin pyridine complex ≥98.0% (by titrimetric analysis)
2,4,6-Trivinylboroxin pyridine complex
CAS Number: 95010-17-6
MDL Number: MFCD03839440
Molecular Formula: C11H14B3NO3
Molecular Weight: 240.67
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 51
Flash Point (°C): 80
Product Categories: Synthetic Organic Chemistry, Vinylation, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: Trivinylcyclotriboroxan - Pyridine Complex
Formula:
C₁₁H₁₄B₃NO₃ MW: 240.67 g/mol Melting Pt: 51 °C Flash Pt: 80 °C |
MDL Number:
MFCD03839440 CAS Number: 95010-17-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...