5-(4-Methylphenyl)-1H-tetrazole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 5-(p-Tolyl)-1H-tetrazole

T2486-5G T2486-25G
TCT2486-5GEA 153.26 CAD
TCT2486-5G TCT2486-25G
5-(4-Methylphenyl)-1H-tetrazole ≥98.0% (by HPLC, titration analysis)
5-(4-Methylphenyl)-1H-tetrazole

CAS Number: 24994-04-5
MDL Number: MFCD01318164
Molecular Formula: C8H8N4
Molecular Weight: 160.18
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 248


Product Categories: Materials Chemistry, Ligands for Functional Metal Complexes
Other Product Names / Synonyms: 5-(4-Methylphenyl)-1H-tetrazole

Formula: C₈H₈N₄
MW: 160.18 g/mol
Melting Pt: 248 °C
MDL Number: MFCD01318164
CAS Number: 24994-04-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70208244916a01e12b65bdf64953d8d0f3677e8b&vid=70208244916a01e12b65bdf64953d8d0f3677e8b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR