Tris(pentafluorophenyl)phosphine ≥95.0%

Supplier: TCI America

T2484-1G T2484-5G
TCT2484-1GEA 86.9 CAD
TCT2484-1G TCT2484-5G
Tris(pentafluorophenyl)phosphine ≥95.0%
Tris(pentafluorophenyl)phosphine

CAS Number: 1259-35-4
MDL Number: MFCD00079654
Molecular Formula: C18F15P
Molecular Weight: 532.15
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 116


Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: Tris(perfluorophenyl)phosphine

Formula: C₁₈F₁₅P
Melting Pt: 116 °C
MDL Number: MFCD00079654
CAS Number: 1259-35-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1831d1f45dfa9dd897cd171537096f3585540009&vid=1831d1f45dfa9dd897cd171537096f3585540009&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR