1-Bromoperfluorohexane ≥97.0%

Supplier: TCI America

T2479-5G T2479-25G
TCT2479-5GEA 146.94 CAD
TCT2479-5G TCT2479-25G
1-Bromoperfluorohexane ≥97.0%
1-Bromoperfluorohexane

CAS Number: 335-56-8
MDL Number: MFCD00042349
Molecular Formula: C6BrF13
Molecular Weight: 398.95
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 100
Specific Gravity (20/20): 1.90


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: 1-Bromotridecafluorohexane, 1-Bromoperfluorohexane, Perfluorohexyl Bromide

Formula: BrF₂C(CF₂)₄CF₃
Boiling Pt: 100 °C
Density: 1.90 (20 °C)
MDL Number: MFCD00042349
CAS Number: 335-56-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f3643834454fee379b49ee49653d24490f7d75d&vid=2d8e63a42bce1457756266bd7b18eeb316a81b57&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR