Triethyl-1,1,2-ethanetricarboxylate ≥96.0%

Supplier: TCI America

T2431-25G T2431-5G
TCT2431-25GEA 100.8 CAD
TCT2431-25G TCT2431-5G
Triethyl-1,1,2-ethanetricarboxylate ≥96.0%
Triethyl 1,1,2-ethanetricarboxylate

CAS Number: 7459-46-3
MDL Number: MFCD00009154
Molecular Formula: C11H18O6
Molecular Weight: 246.26
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 165
Flash Point (°C): 109
Specific Gravity (20/20): 1.09


Other Product Names / Synonyms: 1,1,2-Ethanetricarboxylic Acid Triethyl Ester, Diethyl 2-(Ethoxycarbonyl)butanedioate, 2-(Ethoxycarbonyl)butanedioic Acid Diethyl Ester

Formula: C₁₁H₁₈O₆
Boiling Pt: 165 °C
Density: 1.09 (20 °C)
Flash Pt: 109 °C
MDL Number: MFCD00009154
CAS Number: 7459-46-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f5a08b97ea745f6cc4c4533b7fa30aaa7445610&vid=2f5a08b97ea745f6cc4c4533b7fa30aaa7445610&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR