4-(Trifluoromethyl)cyclohexanecarboxylic acid (cis and trans mixture) ≥98.0%

Supplier: TCI America

Synonyms: 4-(Trifluoromethyl)cyclohexane-1-carboxylic acid

T2417-5G T2417-1G
TCT2417-5GEA 109.02 CAD
TCT2417-5G TCT2417-1G
4-(Trifluoromethyl)cyclohexanecarboxylic acid (cis and trans mixture) ≥98.0%
4-(Trifluoromethyl)cyclohexanecarboxylic acid (cis and trans mixture)

CAS Number: 95233-30-0
MDL Number: MFCD00220049
Molecular Formula: C8H11F3O2
Molecular Weight: 196.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 75

Formula: C₈H₁₁F₃O₂
MW: 196.17 g/mol
Melting Pt: 75 °C
MDL Number: MFCD00220049
CAS Number: 95233-30-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca6135b1771a7803639fdf5f44d9fb04c3d328c9&vid=ca6135b1771a7803639fdf5f44d9fb04c3d328c9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR