2-(p-Toluenesulfonyl)acetic acid ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
2-Tosylacetic acid
T2416-25G
T2416-5G
TCT2416-25GEA
775.78
CAD
TCT2416-25G
TCT2416-5G
2-(p-Toluenesulfonyl)acetic acid ≥98.0% (by HPLC)
2-(p-Toluenesulfonyl)acetic acid
CAS Number: 3937-96-0
MDL Number: MFCD00021764
Molecular Formula: C9H10O4S
Molecular Weight: 214.24
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 118
Formula:
C₉H₁₀O₄S MW: 214.24 g/mol Melting Pt: 118 °C |
MDL Number:
MFCD00021764 CAS Number: 3937-96-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...