4-(Trifluoromethylthio)benzaldehyde ≥90.0%
Supplier: TCI America
T2405-25G
T2405-5G
TCT2405-25GEA
1003.3
CAD
TCT2405-25G
TCT2405-5G
4-(Trifluoromethylthio)benzaldehyde ≥90.0%
4-(Trifluoromethylthio)benzaldehyde
CAS Number: 4021-50-5
MDL Number: MFCD00129191
Molecular Formula: C8H5F3OS
Molecular Weight: 206.18
Purity/Analysis Method: >90.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 85
Flash Point (°C): 100
Freezing point (°C): 26
Formula:
C₈H₅F₃OS Boiling Pt: 85 °C Flash Pt: 100 °C |
MDL Number:
MFCD00129191 CAS Number: 4021-50-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...