4-((Trifluoromethyl)thio)benzoic acid ≥97.0%

Supplier: TCI America

T2390-5G T2390-25G
TCT2390-5GEA 100.8 CAD
TCT2390-5G TCT2390-25G
4-((Trifluoromethyl)thio)benzoic acid ≥97.0%
4-((Trifluoromethyl)thio)benzoic acid

CAS Number: 330-17-6
MDL Number: MFCD00040906
Molecular Formula: C8H5F3O2S
Molecular Weight: 222.18
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 161

Formula: C₈H₅F₃O₂S
MW: 222.19 g/mol
Melting Pt: 161 °C
MDL Number: MFCD00040906
CAS Number: 330-17-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1af8e1bef438d80aa8eb3ad982ed285e3bd464a4&vid=1af8e1bef438d80aa8eb3ad982ed285e3bd464a4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR