2,3,5-Trifluorobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

T2385-5G T2385-1G
TCT2385-5GEA 567.22 CAD
TCT2385-5G TCT2385-1G
2,3,5-Trifluorobenzoic acid ≥98.0% (by GC, titration analysis)
2,3,5-Trifluorobenzoic acid

CAS Number: 654-87-5
MDL Number: MFCD00061200
Molecular Formula: C7H3F3O2
Molecular Weight: 176.09
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 109

Formula: C₇H₃F₃O₂
Melting Pt: 109 °C
MDL Number: MFCD00061200
CAS Number: 654-87-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5549735fc90596dc814a07fab04ae9cc32291157&vid=5549735fc90596dc814a07fab04ae9cc32291157&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR