3-(Trifluoromethoxy)benzoic acid ≥98.0%

Supplier: TCI America

T2383-1G T2383-5G
TCT2383-1GEA 62.44 CAD
TCT2383-1G TCT2383-5G
3-(Trifluoromethoxy)benzoic acid ≥98.0%
3-(Trifluoromethoxy)benzoic acid

CAS Number: 1014-81-9
MDL Number: MFCD00041501
Molecular Formula: C8H5F3O3
Molecular Weight: 206.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 92


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: alpha,alpha,alpha-Trifluoro-m-anisic Acid

Formula: C₈H₅F₃O₃
MW: 206.12 g/mol
Boiling Pt: 272
Melting Pt: 92 °C
MDL Number: MFCD00041501
CAS Number: 1014-81-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098329791dfeafece859c8a4a404169c35ec378b&vid=c1e8cdf4eeeab3263af9a8e353d75fa3b8bfc8df&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR