Tetrafluorophthalic anhydride ≥95.0% (by titrimetric analysis)
Supplier: TCI America
T2381-1G
T2381-5G
TCT2381-1GEA
66.36
CAD
TCT2381-1G
TCT2381-5G
Tetrafluorophthalic anhydride ≥95.0% (by titrimetric analysis)
Tetrafluorophthalic anhydride
CAS Number: 652-12-0
MDL Number: MFCD00039697
Molecular Formula: C8F4O3
Molecular Weight: 220.08
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Color: White
Melting point (°C): 94
Formula:
C₈F₄O₃ Melting Pt: 94 °C |
MDL Number:
MFCD00039697 CAS Number: 652-12-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...