4-Pyridineboronic acid pinacol ester ≥98.0%

Supplier: TCI America

Synonyms: Pyridine-4-boronic acid pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

T2349-1G T2349-5G
TCT2349-1GEA 55.3 CAD
TCT2349-1G TCT2349-5G
4-Pyridineboronic acid pinacol ester ≥98.0%
4-Pyridineboronic acid pinacol ester

CAS Number: 181219-01-2
MDL Number: MFCD01319051
Molecular Formula: C11H16BNO2
Molecular Weight: 205.06
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 151


Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: 2-(4-Pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Pyridylboronic Acid Pinacol Ester

Formula: C₁₁H₁₆BNO₂
MW: 205.06 g/mol
Melting Pt: 151 °C
MDL Number: MFCD01319051
CAS Number: 181219-01-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78ea68046f1a97970a75a0f019fffdb7fbd9989b&vid=78ea68046f1a97970a75a0f019fffdb7fbd9989b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR