3,4,5-Trifluorobenzaldehyde ≥98.0%
Supplier: TCI America
T2321-5G
T2321-1G
TCT2321-5GEA
146.94
CAD
TCT2321-5G
TCT2321-1G
3,4,5-Trifluorobenzaldehyde ≥98.0%
3,4,5-Trifluorobenzaldehyde
CAS Number: 132123-54-7
MDL Number: MFCD00083523
Molecular Formula: C7H3F3O
Molecular Weight: 160.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 174
Flash Point (°C): 110
Freezing point (°C): 28
Formula:
C₇H₃F₃O Boiling Pt: 174 °C Flash Pt: 110 °C |
MDL Number:
MFCD00083523 CAS Number: 132123-54-7 UN: 2811 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...