2,4,6-Trifluorobenzaldehyde ≥98.0%

Supplier: TCI America

T2320-1G T2320-5G
TCT2320-1GEA 72.68 CAD
TCT2320-1G TCT2320-5G
2,4,6-Trifluorobenzaldehyde ≥98.0%
2,4,6-Trifluorobenzaldehyde

CAS Number: 58551-83-0
MDL Number: MFCD00061197
Molecular Formula: C7H3F3O
Molecular Weight: 160.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 66

Formula: C₇H₃F₃O
Melting Pt: 66 °C
MDL Number: MFCD00061197
CAS Number: 58551-83-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=86d072535cd9e9c499eac950d274b33bedb3ac51&vid=86d072535cd9e9c499eac950d274b33bedb3ac51&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR