2,3,6-Trifluorobenzaldehyde ≥98.0%

Supplier: TCI America

T2318-1G T2318-5G
TCT2318-1GEA 47.87 CAD
TCT2318-1G TCT2318-5G
2,3,6-Trifluorobenzaldehyde ≥98.0%
2,3,6-Trifluorobenzaldehyde

CAS Number: 104451-70-9
MDL Number: MFCD00061195
Molecular Formula: C7H3F3O
Molecular Weight: 160.10
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 159
Flash Point (°C): 76
Specific Gravity (20/20): 1.44

Formula: C₇H₃F₃O
MW: 160.1 g/mol
Boiling Pt: 159 °C
Density: 1.44 (20 °C)
Flash Pt: 76 °C
MDL Number: MFCD00061195
CAS Number: 104451-70-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f201dcf23284f41431bef4697b3ddc8ece961332&vid=f201dcf23284f41431bef4697b3ddc8ece961332&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR