Tris(pentafluorophenyl)borane ≥97.0% (by NMR)

Supplier: TCI America

T2313-1G T2313-5G
TCT2313-1GEA 303.83 CAD
TCT2313-1G TCT2313-5G
Tris(pentafluorophenyl)borane ≥97.0% (by NMR)
Tris(pentafluorophenyl)borane

CAS Number: 1109-15-5
MDL Number: MFCD00269813
Molecular Formula: C18BF15
Molecular Weight: 511.98
Purity/Analysis Method: >97.0% (NMR)
Form: Crystal
Color: White
Melting point (°C): 128
Lambda max.: 306 nm (Toluene)

Formula: C₁₈BF₁₅
Melting Pt: 128 °C
MDL Number: MFCD00269813
CAS Number: 1109-15-5
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd0e5824a4c40cbe7793af157106b4548f98a939&vid=dd0e5824a4c40cbe7793af157106b4548f98a939&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (NMR)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR