2,2,3,3-Tetrafluoro-1,4-butanediol ≥95.0%
Supplier: TCI America
Synonyms:
2,2,3,3-Tetrafluorobutane-1,4-diol
T2296-1G
T2296-5G
T2296-25G
TCT2296-1GEA
39.5
CAD
TCT2296-1G
TCT2296-5G
TCT2296-25G
2,2,3,3-Tetrafluoro-1,4-butanediol ≥95.0%
2,2,3,3-Tetrafluoro-1,4-butanediol
CAS Number: 425-61-6
Molecular Formula: C4H6F4O2
Molecular Weight: 162.08
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 112
Melting point (°C): 80
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,4-Dihydroxy-2,2,3,3-tetrafluorobutane
Formula:
C₄H₆F₄O₂ MW: 162.08 g/mol Boiling Pt: 112 °C Melting Pt: 80 °C |
MDL Number:
MFCD00042375 CAS Number: 425-61-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...