Trimethyl orthobenzoate ≥95.0%

Supplier: TCI America

T2287-25G T2287-5G
TCT2287-25GEA 86.9 CAD
TCT2287-25G TCT2287-5G
Trimethyl orthobenzoate ≥95.0%
Trimethyl orthobenzoate

CAS Number: 707-07-3
MDL Number: MFCD00008474
Molecular Formula: C10H14O3
Molecular Weight: 182.22
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 115
Flash Point (°C): 82
Specific Gravity (20/20): 1.07


Other Product Names / Synonyms: Orthobenzoic Acid Trimethyl Ester, alpha,alpha,alpha-Trimethoxytoluene, Methyl Orthobenzoate

Formula: C₁₀H₁₄O₃
Boiling Pt: 115 °C
Density: 1.07 (20 °C)
Flash Pt: 82 °C
MDL Number: MFCD00008474
CAS Number: 707-07-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68e463574c61de7fb49d27f3340bddfa9ef2f9cd&vid=68e463574c61de7fb49d27f3340bddfa9ef2f9cd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR