3,4,5-Trichloro-α,α,α-trifluorotoluene ≥98.0%

Supplier: TCI America

T2281-25G T2281-5G
TCT2281-25GEA 61.62 CAD
TCT2281-25G TCT2281-5G
3,4,5-Trichloro-α,α,α-trifluorotoluene ≥98.0%
3,4,5-Trichloro-α,α,α-trifluorotoluene

CAS Number: 50594-82-6
Molecular Formula: C7H2Cl3F3
Molecular Weight: 249.44
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 113
Melting point (°C): -6
Flash Point (°C): 98
Specific Gravity (20/20): 1.60


Other Product Names / Synonyms: 1,2,3-Trichloro-5-(trifluoromethyl)benzene

Formula: C₇H₂Cl₃F₃
Boiling Pt: 113 °C
Melting Pt: –6 °C
Density: 1.60 (20 °C)
Flash Pt: 98 °C
MDL Number: MFCD00035985
CAS Number: 50594-82-6
UN: 2234
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c902f92963b7036d6c91d334eabd8947d0aa24fe&vid=c902f92963b7036d6c91d334eabd8947d0aa24fe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR