4-(Trifluoromethoxy)benzaldehyde ≥97.0%

Supplier: TCI America

T2256-25G T2256-5G
TCT2256-25GEA 273.34 CAD
TCT2256-25G TCT2256-5G
4-(Trifluoromethoxy)benzaldehyde ≥97.0%
4-(Trifluoromethoxy)benzaldehyde

CAS Number: 659-28-9
Molecular Formula: C8H5F3O2
Molecular Weight: 190.12
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 93
Flash Point (°C): 70
Specific Gravity (20/20): 1.33

Formula: C₈H₅F₃O₂
Boiling Pt: 93 °C
Density: 1.33 (20 °C)
Flash Pt: 70 °C
MDL Number: MFCD00041530
CAS Number: 659-28-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a052480042b6ba99f51c2c3f5bd6da116ba1a70&vid=7a052480042b6ba99f51c2c3f5bd6da116ba1a70&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR