Tetrahydrofurfuryl acrylate ≥98.0% stabilised

Supplier: TCI America

Synonyms: (Tetrahydrofuran-2-yl)methyl acrylate, Acrylic Acid Tetrahydrofurfuryl Ester

T2239-25G T2239-500G
TCT2239-25GEA 60.04 CAD
TCT2239-25G TCT2239-500G
Tetrahydrofurfuryl acrylate ≥98.0% stabilised
Tetrahydrofurfuryl acrylate

(stabilized with MEHQ)
CAS Number: 2399-48-6
Molecular Formula: C8H12O3
Molecular Weight: 156.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 87
Flash Point (°C): 95
Specific Gravity (20/20): 1.07


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Acrylic Acid Tetrahydrofurfuryl Ester

Formula: C₈H₁₂O₃
Boiling Pt: 87 °C
Density: 1.07 (20 °C)
Flash Pt: 95 °C
MDL Number: MFCD00014516
CAS Number: 2399-48-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d85916d78d2a9c8e0fd027771bb18aee50d682fd&vid=d85916d78d2a9c8e0fd027771bb18aee50d682fd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR