1-Tritylimidazole-4-carbaldehyde ≥98.0%

Supplier: TCI America

Synonyms: 1-Trityl-1H-imidazole-4-carboxaldehyde, 1-Trityl-1H-imidazole-4-carbaldehyde, 1-Tritylimidazole-4-carboxaldehyde

T2204-100MG T2204-1G
TCT2204-100MGEA 33.18 CAD
TCT2204-100MG TCT2204-1G
1-Tritylimidazole-4-carbaldehyde ≥98.0%
1-Tritylimidazole-4-carbaldehyde

CAS Number: 33016-47-6
Molecular Formula: C23H18N2O
Molecular Weight: 338.41
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 186


Other Product Names / Synonyms: 4-Formyl-1-tritylimidazole, 1-(Triphenylmethyl)imidazole-4-carboxaldehyde

Formula: C₂₃H₁₈N₂O
MW: 338.41 g/mol
Melting Pt: 186 °C
MDL Number: MFCD02179554
CAS Number: 33016-47-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c4aec114c4f3718ca2a9e24bd11bb418cc7efbb&vid=8c4aec114c4f3718ca2a9e24bd11bb418cc7efbb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR