2,3,4,5-Tetraphenylthiophene ≥98.0%

Supplier: TCI America

Synonyms: Tetraphenylthiophene

T2200-5G T2200-25G
TCT2200-5GEA 66.36 CAD
TCT2200-5G TCT2200-25G
2,3,4,5-Tetraphenylthiophene ≥98.0%
2,3,4,5-Tetraphenylthiophene

CAS Number: 1884-68-0
MDL Number: MFCD00014527
Molecular Formula: C28H20S
Molecular Weight: 388.53
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 186

Formula: C₂₈H₂₀S
MW: 388.53 g/mol
Melting Pt: 186 °C
MDL Number: MFCD00014527
CAS Number: 1884-68-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=baf75e98a107a2b58a380529553864aa76049f0e&vid=baf75e98a107a2b58a380529553864aa76049f0e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR