Tris(dibenzylideneacetone)dipalladium(0) ≥75.0% (by titrimetric analysis)

Supplier: TCI America

T2184-5G T2184-1G
TCT2184-5GEA 1045.96 CAD
TCT2184-5G TCT2184-1G
Tris(dibenzylideneacetone)dipalladium(0) ≥75.0% (by titrimetric analysis)
Tris(dibenzylideneacetone)dipalladium(0)

CAS Number: 51364-51-3
Molecular Formula: C51H42O3Pd2
Molecular Weight: 915.73
Purity/Analysis Method: >75.0% (T)
Form: Crystal
Color: Deep Yellow Red
Melting point (°C): 155

Formula: C₅₁H₄₂O₃Pd₂
Melting Pt: 155 °C
MDL Number: MFCD00013310
CAS Number: 51364-51-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7bd1738d3d3e0a280f4251d3de861eb819d49bee&vid=7bd1738d3d3e0a280f4251d3de861eb819d49bee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 75.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR