Ruthenium(III) acetylacetonate
Supplier: TCI America
Synonyms:
Ruthenium (III) 2,4-pentanedionate, Tris(2,4-pentanedionato-O,O')ruthenium (III), Ruthenium (III) tris(2,4-pentanedionato-O,O'), Tris(2,4-pentanedionato)ruthenium(III)
T2183-1G
T2183-5G
TCT2183-1GEA
148.33
CAD
TCT2183-1G
TCT2183-5G
Ruthenium(III) acetylacetonate
Ruthenium (III) acetylacetonate
CAS Number: 14284-93-6
MDL Number: MFCD00000030
Molecular Formula: C15H21O6Ru
Molecular Weight: 398.40
Form: Crystal
Color: Deep Red
Melting point (°C): 260
Other Product Names / Synonyms: Acetylacetone Ruthenium(III) Salt, Ruthenium(III) Acetylacetonate
Formula:
C₁₅H₂₁O₆Ru MW: 398.4 g/mol Melting Pt: 260 °C |
MDL Number:
MFCD00000030 CAS Number: 14284-93-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...