2,3,4-Trihydroxybenzaldehyde ≥98.0%

Supplier: TCI America

T2158-25G T2158-5G
TCT2158-25GEA 139.04 CAD
TCT2158-25G TCT2158-5G
2,3,4-Trihydroxybenzaldehyde ≥98.0%
2,3,4-Trihydroxybenzaldehyde

CAS Number: 2144-08-3
Molecular Formula: C7H6O4
Molecular Weight: 154.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 162


Other Product Names / Synonyms: Pyrogallol-4-carboxaldehyde

Formula: C₇H₆O₄
MW: 154.12
Boiling Pt: 332
Melting Pt: 159
Density: 1.054
MDL Number: MFCD00003325
CAS Number: 2144-08-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d42282fb109c4eb69e7eabcbd98a36b5a09c24b3&vid=e5e530fe11e5bc4037c6fa87fa42ed0f9666d8fd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR