1-Nitro-4-(trifluoromethoxy)benzene ≥96.0%

Supplier: TCI America

Synonyms: 4-(Trifluoromethoxy)nitrobenzene

T2155-5G T2155-25G
TCT2155-5GEA 26.86 CAD
TCT2155-5G TCT2155-25G
1-Nitro-4-(trifluoromethoxy)benzene ≥96.0%
1-Nitro-4-(trifluoromethoxy)benzene

CAS Number: 713-65-5
MDL Number: MFCD00040302
Molecular Formula: C7H4F3NO3
Molecular Weight: 207.11
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Color: Slightly Pale Yellow
Boiling point (°C): 60
Melting point (°C): 15
Specific Gravity (20/20): 1.45

Formula: C₇H₄F₃NO₃
MW: 207.11 g/mol
Boiling Pt: 60 °C
Melting Pt: 15 °C
Density: 1.45 (20 °C)
MDL Number: MFCD00040302
CAS Number: 713-65-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=892167a7263fb92ca5ceee214f4537a96aca693c&vid=892167a7263fb92ca5ceee214f4537a96aca693c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR