1H,1H,2H,2H-Perfluoro-1-iodooctane ≥97.0%

Supplier: TCI America

T2074-5G T2074-25G
TCT2074-5GEA 54.67 CAD
TCT2074-5G TCT2074-25G
1H,1H,2H,2H-Perfluoro-1-iodooctane ≥97.0%
1H,1H,2H,2H-Perfluoro-1-iodooctane

CAS Number: 2043-57-4
MDL Number: MFCD00039410
Molecular Formula: C8H4F13I
Molecular Weight: 474.00
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 180
Flash Point (°C): 150
Specific Gravity (20/20): 1.95


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: 1H,1H,2H,2H-Perfluoro-n-octyl Iodide, 1H,1H,2H,2H-Tridecafluoro-1-iodo-n-octane, 1H,1H,2H,2H-Perfluoro-1-iodo-n-octane

Formula: C₈H₄F₁₃I
Boiling Pt: 180 °C
Density: 1.95 (20 °C)
Flash Pt: 150 °C
MDL Number: MFCD00039410
CAS Number: 2043-57-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=993951b1984d758598136a387c75c2cd1d650bd7&vid=993951b1984d758598136a387c75c2cd1d650bd7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR