2,3,4-Trimethoxycinnamic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-(2,3,4-Trimethoxyphenyl)acrylic acid

T2016-5G T2016-25G
TCT2016-5GEA 112.34 CAD
TCT2016-5G TCT2016-25G
2,3,4-Trimethoxycinnamic acid ≥98.0% (by GC, titration analysis)
2,3,4-Trimethoxycinnamic acid

CAS Number: 33130-03-9
MDL Number: MFCD00014376
Molecular Formula: C12H14O5
Molecular Weight: 238.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 173


Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: 3-(2,3,4-Trimethoxyphenyl)acrylic Acid

Formula: C₁₂H₁₄O₅
MW: 238.24 g/mol
Melting Pt: 173 °C
MDL Number: MFCD00014376
CAS Number: 33130-03-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8d2071053437bd8d9b51e29608182395ada21d8&vid=f8d2071053437bd8d9b51e29608182395ada21d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR